Learn More
Medchemexpress LLC (Z)-Semaxanib (SU5416) | 194413-58-6 | MFCD09763655 | >99.0% | 238.29 g/mol | C15H14N2O | 1 G
SDP
Catalog No. 5000247737
Encompass_Preferred
(Z)-Semaxanib is a small-molecule inhibitor of vascular endothelial growth factor receptors (VEGFR) and related protein tyrosine kinases, used as a research tool to study angiogenesis, endothelial cell proliferation, and tumor biology. The compound is provided as a powder and requires appropriate low-temperature storage and handling.
  • Inhibits VEGFR and related tyrosine kinases.
  • Shows cytotoxicity in various cell lines (reported IC50 values in the micromolar range).
  • Suitable for in vitro and in vivo research applications.
  • Supplied as a powder with recommended cold storage.
  • Available in multiple pack sizes, including 1 g.
Catalog No. 50-002-47737 Supplier Medchemexpress LLC Supplier No. HY103321G
(Z)-Semaxanib is a small-molecule inhibitor of vascular endothelial growth factor receptors (VEGFR) and related protein tyrosine kinases, used as a research tool to study angiogenesis, endothelial cell proliferation, and tumor biology. The compound is provided as a powder and requires appropriate low-temperature storage and handling.
  • Inhibits VEGFR and related tyrosine kinases.
  • Shows cytotoxicity in various cell lines (reported IC50 values in the micromolar range).
  • Suitable for in vitro and in vivo research applications.
  • Supplied as a powder with recommended cold storage.
  • Available in multiple pack sizes, including 1 g.