Learn More
Medchemexpress LLC Z-Ser-obzl | 21209-51-8 | MFCD00037824 | ≥97.0% | 329.35 g/mol | C18H19NO5 | 10 G
SDP
Catalog No. 5000210080
Encompass_Preferred
Z-Ser-obzl (CAS 21209-51-8) is a CBZ-protected L-serine benzyl ester used as a biochemical reagent and protected amino-acid building block for peptide synthesis and organic synthesis research.
  • Used as a protected amino-acid building block for peptide synthesis.
  • Commonly used as an organic intermediate in medicinal chemistry and peptide research.
  • Typical purity ≥97.0% (HPLC).
  • Molecular formula C18H19NO5; molecular weight 329.35 g/mol.
  • Supplied in small laboratory pack sizes; verify exact pack size and certificates with the supplier before use.
Catalog No. 50-002-10080 Supplier Medchemexpress LLC Supplier No. HYW04400110G
Z-Ser-obzl (CAS 21209-51-8) is a CBZ-protected L-serine benzyl ester used as a biochemical reagent and protected amino-acid building block for peptide synthesis and organic synthesis research.
  • Used as a protected amino-acid building block for peptide synthesis.
  • Commonly used as an organic intermediate in medicinal chemistry and peptide research.
  • Typical purity ≥97.0% (HPLC).
  • Molecular formula C18H19NO5; molecular weight 329.35 g/mol.
  • Supplied in small laboratory pack sizes; verify exact pack size and certificates with the supplier before use.