Learn More
Beckman Coulter Z-SERIES TRI PACK
Catalog No. NC9647749
Encompass
Z Series Tri-Pak; Mercury Free, compact Particle Counters/Size Analyzers, utilizing Coulter Principle (Electrical Sensing Zone Method) for cell counting, particle counting, size analysis
Catalog No. NC9647749 Supplier Beckman Coulter Supplier No. 8320312
Z Series Tri-Pak; Mercury Free, compact Particle Counters/Size Analyzers, utilizing Coulter Principle (Electrical Sensing Zone Method) for cell counting, particle counting, size analysis