Learn More
TARGETMOL CHEMICALS INC Z-Stat 1MG
SDP
Catalog No. 502878728
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. ζ-Stat (NSC37044), a specific and atypical inhibitor of PKC-ζ with an IC50 of 5 uM, demonstrates the ability to inhibit proliferation and induce apoptosis in melanoma cell lines, exhibiting antitumor activity in vitro[1][2]. Purity 98.01%
Catalog No. 50-287-8728 Supplier TARGETMOL CHEMICALS INC Supplier No. T354071MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. ζ-Stat (NSC37044), a specific and atypical inhibitor of PKC-ζ with an IC50 of 5 uM, demonstrates the ability to inhibit proliferation and induce apoptosis in melanoma cell lines, exhibiting antitumor activity in vitro[1][2]. Purity 98.01%