Learn More
Medchemexpress LLC Z-Tyr-Lys-Arg-pNA | 108318-36-1 | 99.90% | 719.79 | 10 MG
SDP
Catalog No. 5000398550
Encompass_Preferred
Z-Tyr-Lys-Arg-pNA is a chromogenic substrate designed for detecting the activity of subtilisin-type and yapsin-like proteases. It is intended for research use only.
  • Chromogenic substrate
  • Detects subtilisin-type protease activity
  • Detects yapsin-like protease activity
  • Soluble in DMSO at 100 mg/mL (138.93 mM)
  • For research use only
Catalog No. 50-003-98550 Supplier Medchemexpress LLC Supplier No. HYP451710MG
Z-Tyr-Lys-Arg-pNA is a chromogenic substrate designed for detecting the activity of subtilisin-type and yapsin-like proteases. It is intended for research use only.
  • Chromogenic substrate
  • Detects subtilisin-type protease activity
  • Detects yapsin-like protease activity
  • Soluble in DMSO at 100 mg/mL (138.93 mM)
  • For research use only