Learn More
CPC Scientific Z-Tyr-Val-Ala-Asp-AFC 1MG
SDP
Supplier:  CPC Scientific CASP034A
Encompass
When halogen substituents at the fluorescent group are present, the membrane permeability of this caspase-1 substrate greatly rises.ONE-LETTER SEQUENCE: Z-YVAD-AFCMOLECULAR FORMULA: C39H40F3N5O11MOLECULAR WEIGHT:811.8STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [201608-13-1], RESEARCH AREA: Apoptosis
Catalog No. 50-210-8980