Learn More
CPC Scientific Z-Tyr-Val-Ala-Asp-CMK 0.5MG
SDP
Supplier:  CPC Scientific CASP069A
Encompass
Z-Tyr-Val-Ala-Asp-CMK is an inhibitor of caspase-1.SEQUENCE: Z-Tyr-Val-Ala-Asp-CMKONE-LETTER SEQUENCE: Z-YVAD-CMKMOLECULAR FORMULA: C30H37Cl1N4O9MOLECULAR WEIGHT: 633.10STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [402474-85-5]SYNONYMS: Z-Tyr-Val-Ala-Asp-chloromethylketoneRESEARCH AREA: ApoptosisREFERENCES:S. Shoma et al., Infect. Immun., 76, 1547 (2008)K. Wallner et al., Arterioscler. Thromb. Vasc. Biol., 24, 1416 (2004)U. E. Knies et al., Proc. Natl. Acad. Sci. USA, 95, 12322 (1998)V. L. Longthorne and G. T. Williams, EMBO J., 16, 3805 (1997)SKU(s): CASP-069A, CASP-069B
Catalog No. 50-210-9045