Learn More
Zebra Technologies Corporation Label, Polyester, 2x1in (50.8x25.4mm); TT, Z-Ultimate 3000T White, High Performance Coated, Permanent Adhesive, 1in (25.4mm) core, 2530/roll, 12/box, Plain
Supplier:  Zebra Technologies Corporation 17157
Encompass_Preferred
This item has a minimum order quantity of 12 per supplier requirements
Label, Polyester, 2x1in (50.8x25.4mm); TT, Z-Ultimate 3000T White, High Performance Coated, Permanent Adhesive, 1in (25.4mm) core, 2530/roll, 12/box, Plain
Catalog No. NC1574128