Learn More
Cayman Chemical Z-VA-DL-D(OMe)-FMK (trifluoroacetate salt), Purity: ≥95%, 1 mg
SDP
Catalog No. 502941039
Encompass_Preferred
A pan-caspase inhibitor; inhibits anti-Fas antibody-induced cleavage of caspase-3 and PARP in Jurkat T lymphocytes at 0.5 μM; inhibits cell death induced by TNF-α and cycloheximide co-application in HeLa cells at 50 μM; inhibits radiation-induced increases in the number of TUNEL-positive neurons in the rat hypoglossal nucleus when administered intracerebroventricularly at 2 μg/animal
Catalog No. 50-294-1039 Supplier Cayman Chemical Supplier No. 274211 MG
A pan-caspase inhibitor; inhibits anti-Fas antibody-induced cleavage of caspase-3 and PARP in Jurkat T lymphocytes at 0.5 μM; inhibits cell death induced by TNF-α and cycloheximide co-application in HeLa cells at 50 μM; inhibits radiation-induced increases in the number of TUNEL-positive neurons in the rat hypoglossal nucleus when administered intracerebroventricularly at 2 μg/animal