Learn More
Medchemexpress LLC Z-VAD | 162852-62-2 | 99.8% | 437.44 | 50 MG
SDP
Catalog No. 5000392149
Encompass_Preferred
Z-VAD is an irreversible, broad-spectrum pan-caspase inhibitor that can inhibit various caspases including caspase-3, -6, -7, -8, -9, with a weaker effect on caspase-2. It can block apoptosis signaling pathways, induce autophagy and necrosis in tumor cells, and exhibits anti-angiogenic activity. It is primarily used in research related to cancer radiosensitization and cell death pathway regulation.
  • Irreversible, broad-spectrum pan-caspase inhibitor
  • Inhibits caspases including caspase-3, -6, -7, -8, and -9
  • Blocks apoptosis signaling pathways
  • Induces autophagy and necrosis in tumor cells
  • Exhibits anti-angiogenic activity
  • Enhances sensitivity of breast and lung cancer cells to radiotherapy
  • Prolongs growth delay of tumor xenograft models
  • Well tolerated in studies
Catalog No. 50-003-92149 Supplier Medchemexpress LLC Supplier No. HY16438850MG
Z-VAD is an irreversible, broad-spectrum pan-caspase inhibitor that can inhibit various caspases including caspase-3, -6, -7, -8, -9, with a weaker effect on caspase-2. It can block apoptosis signaling pathways, induce autophagy and necrosis in tumor cells, and exhibits anti-angiogenic activity. It is primarily used in research related to cancer radiosensitization and cell death pathway regulation.
  • Irreversible, broad-spectrum pan-caspase inhibitor
  • Inhibits caspases including caspase-3, -6, -7, -8, and -9
  • Blocks apoptosis signaling pathways
  • Induces autophagy and necrosis in tumor cells
  • Exhibits anti-angiogenic activity
  • Enhances sensitivity of breast and lung cancer cells to radiotherapy
  • Prolongs growth delay of tumor xenograft models
  • Well tolerated in studies