Learn More
Cayman Chemical Z-VAD-AMCacetate 500ug
SDP
Catalog No. 502938343
Encompass_Preferred
A caspase-1 fluorogenic substrate; upon enzymatic cleavage, AMC is released and its fluorescence can be used to quantify enzyme activity; ex/em = 340-360/440-460 nm, respectively
Catalog No. 50-293-8343 Supplier Cayman Chemical Supplier No. 27142500 UG
A caspase-1 fluorogenic substrate; upon enzymatic cleavage, AMC is released and its fluorescence can be used to quantify enzyme activity; ex/em = 340-360/440-460 nm, respectively