Learn More
Cayman Chemical Z-VAD-AMC (acetate), Purity: ≥95%, 5 mg
SDP
Catalog No. 502938781
Encompass_Preferred
A caspase-1 fluorogenic substrate; upon enzymatic cleavage, AMC is released and its fluorescence can be used to quantify enzyme activity; ex/em = 340-360/440-460 nm, respectively
Catalog No. 50-293-8781 Supplier Cayman Chemical Supplier No. 271425 MG
A caspase-1 fluorogenic substrate; upon enzymatic cleavage, AMC is released and its fluorescence can be used to quantify enzyme activity; ex/em = 340-360/440-460 nm, respectively