Learn More
Selleck Chemical LLC Z-VAD(OH)-FMK (Caspase Inhibitor VI) 1mg 161401-82-7
SDP
Supplier:  Selleck Chemical LLC S81021MG
Encompass_Preferred
Z-VAD-FMK (Caspase Inhibitor VI) is an irreversible pan-caspase inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1190993