Learn More
Medchemexpress LLC HY-16658B 10mg , Z-VAD-FMK CAS:161401-82-7 Purity:>98%
SDP
Catalog No. 502258982
Encompass_Preferred
HY-16658B 10mg Z-VAD-FMK CAS: 161401-82-7 Z-VAD-FMK (Z-VAD(OH)-FMK) is a well-know pan caspase inhibitor, which does not inhibit ubiquitin carboxy-terminal hydrolase L1 (UCHL1) activity even at concentrations as high as 440 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-8982 Supplier Medchemexpress LLC Supplier No. HY16658B10MG
HY-16658B 10mg Z-VAD-FMK CAS: 161401-82-7 Z-VAD-FMK (Z-VAD(OH)-FMK) is a well-know pan caspase inhibitor, which does not inhibit ubiquitin carboxy-terminal hydrolase L1 (UCHL1) activity even at concentrations as high as 440 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.