Learn More
Apexbio Technology LLC Z-VAD-FMK, 10mM (in 1mL DMSO) Cas No: 187389-52-2 MFCD: MFCD02684037
SDP
Supplier:  Apexbio Technology LLC A19025.1
Encompass_Preferred
Z-VAD-FMK, an inhibitor of ICE-like proteases, inhibits apoptosis in THP.1 cells induced by diverse stimuli1 and Fas antigen-induced apoptosis in Jurkat T-cells2. There are other sizes available. Please inqury us for quote.
Catalog No. NC1498763