Learn More
Invivogen Pan-Caspase inhibitor - Z-VAD-FMK, 1 mg
SDP
Catalog No. NC0118651
Encompass
Z-VAD-FMK is a cell-permeable pan-caspase inhibitor. It potently inhibits human caspase-1 to -10 with the exception of caspase-2. It also inhibits murine caspases, notably caspase-1, caspase-3, and caspase-11, the ortholog of human caspase-4 and -5. Caspases are a family of cysteine proteases that are centrally involved in cell death and inflammation responses.
Catalog No. NC0118651 Supplier Invivogen Supplier No. TLRLVAD
Z-VAD-FMK is a cell-permeable pan-caspase inhibitor. It potently inhibits human caspase-1 to -10 with the exception of caspase-2. It also inhibits murine caspases, notably caspase-1, caspase-3, and caspase-11, the ortholog of human caspase-4 and -5. Caspases are a family of cysteine proteases that are centrally involved in cell death and inflammation responses.