Learn More
Selleck Chemical LLC Z-VAD(OH)-FMK (Caspase Inhibitor VI) 5mg 161401-82-7
SDP
Catalog No. NC1201027
Encompass_Preferred
Z-VAD-FMK (Caspase Inhibitor VI) is an irreversible pan-caspase inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1201027 Supplier Selleck Chemical LLC Supplier No. S81025MG
Z-VAD-FMK (Caspase Inhibitor VI) is an irreversible pan-caspase inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.