Learn More
Medchemexpress LLC HY-16658B 5mg Medchemexpress, Z-VAD-FMK CAS:161401-82-7 Purity:>98%
SDP
Catalog No. 501969975
Encompass_Preferred
Medchemexpress, HY-16658B 5mg Z-VAD-FMK CAS:161401-82-7 Z-VAD-FMK (Z-VAD(OH)-FMK) is a well-know pan caspase inhibitor, which does not inhibit ubiquitin carboxy-terminal hydrolase L1 (UCHL1) activity even at concentrations as high as 440 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9975 Supplier Medchemexpress LLC Supplier No. HY16658B5MG
Medchemexpress, HY-16658B 5mg Z-VAD-FMK CAS:161401-82-7 Z-VAD-FMK (Z-VAD(OH)-FMK) is a well-know pan caspase inhibitor, which does not inhibit ubiquitin carboxy-terminal hydrolase L1 (UCHL1) activity even at concentrations as high as 440 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.