Learn More
Sigma Aldrich Fine Chemicals Biosciences Z VAD FMK solid2MG
Catalog No. 502907566
Encompass
Z-VAD-FMK inhibits yeast and mammalian peptide N-glycanase (PNGase). It induces apoptosis. Z-VAD-FMK might be associated with endoplasmic reticulum (ER) stress.
Catalog No. 50-290-7566 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. V1162MG
Z-VAD-FMK inhibits yeast and mammalian peptide N-glycanase (PNGase). It induces apoptosis. Z-VAD-FMK might be associated with endoplasmic reticulum (ER) stress.