Learn More
Cayman Chemical Z-VAD(OH)-FMK, Purity: ≥95%, CAS: 161401-82-7, 1 mg
SDP
Catalog No. 502971701
Encompass_Preferred
An irreversible tripeptide inhibitor of all caspases; useful in studies involving recombinant, isolated, or purified enzymes
Catalog No. 50-297-1701 Supplier Cayman Chemical Supplier No. 144671 MG
An irreversible tripeptide inhibitor of all caspases; useful in studies involving recombinant, isolated, or purified enzymes