Learn More
Cayman Chemical Z-VAD OH-FMK 500ug
SDP
Catalog No. 502970298
Encompass_Preferred
An irreversible tripeptide inhibitor of all caspases; useful in studies involving recombinant, isolated, or purified enzymes
Catalog No. 50-297-0298 Supplier Cayman Chemical Supplier No. 14467500 UG
An irreversible tripeptide inhibitor of all caspases; useful in studies involving recombinant, isolated, or purified enzymes