Learn More
Cayman Chemical Z-VAD(OH)-FMK, Purity: ≥95%, CAS: 161401-82-7, 5 mg
SDP
Catalog No. 502971584
Encompass_Preferred
An irreversible tripeptide inhibitor of all caspases; useful in studies involving recombinant, isolated, or purified enzymes
Catalog No. 50-297-1584 Supplier Cayman Chemical Supplier No. 144675 MG
An irreversible tripeptide inhibitor of all caspases; useful in studies involving recombinant, isolated, or purified enzymes