Learn More
Medchemexpress LLC HY-16658 1mg Medchemexpress, Z-VAD(OMe)-FMK CAS:187389-52-2 Purity:>98%
SDP
Catalog No. 502029209
Encompass_Preferred
Medchemexpress, HY-16658 1mg Z-VAD(OMe)-FMK CAS:187389-52-2 Z-VAD(OMe)-FMK (Z-Val-Ala-Asp(OMe)-FMK) is a cell-permeable and irreversible pan-caspase inhibitor. Z-VAD(OMe)-FMK is an ubiquitin carboxy-terminal hydrolase L1 (UCHL1) inhibitor. Z-VAD(OMe)-FMK irreversibly modifies UCHL1 by targeting the active site of UCHL1[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9209 Supplier Medchemexpress LLC Supplier No. HY166581MG
Medchemexpress, HY-16658 1mg Z-VAD(OMe)-FMK CAS:187389-52-2 Z-VAD(OMe)-FMK (Z-Val-Ala-Asp(OMe)-FMK) is a cell-permeable and irreversible pan-caspase inhibitor. Z-VAD(OMe)-FMK is an ubiquitin carboxy-terminal hydrolase L1 (UCHL1) inhibitor. Z-VAD(OMe)-FMK irreversibly modifies UCHL1 by targeting the active site of UCHL1[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.