Learn More
Cayman Chemical Z-VAD(OMe)-FMK, Purity: ≥95%, CAS: 187389-52-2, 5 mg
SDP
Catalog No. 502971534
Encompass_Preferred
A cell-permeable, competitive, and irreversible inhibitor of all caspases, inhibiting cleavage of PARP and preventing apoptosis when used at 10-50 µM; blocks caspase-mediated apoptosis in vivo; effectively prevents caspase action in inflammasomes
Catalog No. 50-297-1534 Supplier Cayman Chemical Supplier No. 144635 MG
A cell-permeable, competitive, and irreversible inhibitor of all caspases, inhibiting cleavage of PARP and preventing apoptosis when used at 10-50 µM; blocks caspase-mediated apoptosis in vivo; effectively prevents caspase action in inflammasomes