Learn More
TARGETMOL CHEMICALS INC Z-VADOME-FMK 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T60135MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Z-VAD(OMe)-FMK (Z-Val-Ala-Asp(OMe)-FMK) is a cell-permeable and irreversible pan-caspase inhibitor. It inhibits cleavage of PARP preventing apoptosis when used at 10-50 uM. purity: 99%
Catalog No. 50-257-9667