Learn More
Medchemexpress LLC Z-Val-Ala-Asp | 162852-62-2 | 99.8% | 437.44 | 25 MG
SDP
Catalog No. 5000391496
Encompass_Preferred
Z-VAD is an irreversible, broad-spectrum pan-caspase inhibitor that can inhibit various caspases including caspase-3, -6, -7, -8, -9. It blocks apoptosis signaling pathways, induces autophagy and necrosis in tumor cells, and exhibits anti-angiogenic activity.
  • Enhances sensitivity of breast cancer and lung cancer cells to radiotherapy.
  • Prolongs growth delay of tumor xenograft models.
  • Well tolerated in research models.
  • Used for cancer radiosensitization and cell death pathway regulation studies.
Catalog No. 50-003-91496 Supplier Medchemexpress LLC Supplier No. 25MGHY164388
Z-VAD is an irreversible, broad-spectrum pan-caspase inhibitor that can inhibit various caspases including caspase-3, -6, -7, -8, -9. It blocks apoptosis signaling pathways, induces autophagy and necrosis in tumor cells, and exhibits anti-angiogenic activity.
  • Enhances sensitivity of breast cancer and lung cancer cells to radiotherapy.
  • Prolongs growth delay of tumor xenograft models.
  • Well tolerated in research models.
  • Used for cancer radiosensitization and cell death pathway regulation studies.