Learn More
CPC Scientific Z-Val-Ala-Asp-AMC 5MG
SDP
Catalog No. 502108983
Encompass
SEQUENCE: Z-Val-Ala-Asp-AMCONE-LETTER SEQUENCE: Z-VAD-AMCMOLECULAR FORMULA: C30H34N4O9MOLECULAR WEIGHT: 594.6STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [219137-91-4]SYNONYMS: RESEARCH AREA: ApoptosisREFERENCES:S.A. Loddick et al., NeuroReport 7, 1465 (1996)SKU(s): CASP-036A, CASP-036B
Catalog No. 50-210-8983 Supplier CPC Scientific Supplier No. CASP036B
SEQUENCE: Z-Val-Ala-Asp-AMCONE-LETTER SEQUENCE: Z-VAD-AMCMOLECULAR FORMULA: C30H34N4O9MOLECULAR WEIGHT: 594.6STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [219137-91-4]SYNONYMS: RESEARCH AREA: ApoptosisREFERENCES:S.A. Loddick et al., NeuroReport 7, 1465 (1996)SKU(s): CASP-036A, CASP-036B