Learn More
Bachem Z-Val-Ala-DL-Asp(OMe)-fluoromethylketone
SDP
Supplier:  Bachem 4027403.0001
Encompass_Preferred
Z-Val-Ala-DL-Asp(OMe)-fluoromethylketone, 634911-81-2, C22H30FN3O7, Mr 467.50, 1mg. Z-VAD(OMe)-FMK, Caspase-1 Inhibitor V. Z-VAD(OMe)-FMK, a pan-caspase inhibitor, is a competitive irreversible peptide inhibitor and blocks caspase-1 family and caspase-3 family enzymes. The methyl ester of Z-VAD-FMK has been used in studies to support the hypothesis that inhibitors of caspases can limit brain infarction resulting from the permanent obstruction of a brain artery.
Catalog No. 50-260-553