Learn More
CPC Scientific Z-Val-Asp-Val-Ala-Asp-AFC 5MG
SDP
Catalog No. 502108999
Encompass
SEQUENCE: Z-Val-Asp-Val-Ala-Asp-AFCONE-LETTER SEQUENCE: Z-VDVAD-AFCMOLECULAR FORMULA: C39H45F3O6O13MOLECULAR WEIGHT: 862.8STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [219138-08-6]SYNONYMS: Caspase-2 substrateRESEARCH AREA: ApoptosisREFERENCES:SKU(s): CASP-045A, CASP-045B
Catalog No. 50-210-8999 Supplier CPC Scientific Supplier No. CASP045B
SEQUENCE: Z-Val-Asp-Val-Ala-Asp-AFCONE-LETTER SEQUENCE: Z-VDVAD-AFCMOLECULAR FORMULA: C39H45F3O6O13MOLECULAR WEIGHT: 862.8STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [219138-08-6]SYNONYMS: Caspase-2 substrateRESEARCH AREA: ApoptosisREFERENCES:SKU(s): CASP-045A, CASP-045B