Learn More
CPC Scientific Z-Val-Asp-Val-Ala-Asp-pNA 5MG
SDP
Supplier:  CPC Scientific CASP046B
Encompass
ONE-LETTER SEQUENCE: ZVDVAD-pNAMOLECULAR FORMULA: C35H45N7O13MOLECULAR WEIGHT:771.8STORAGE CONDITIONS: -20 5CRESEARCH AREA: Apoptosis
Catalog No. 50-210-9001