Learn More
CPC Scientific Z-Val-Glu-Ile-Asp-AFC 25MG
SDP
Supplier:  CPC Scientific CASP074C
Encompass
ONE-LETTER SEQUENCE: ZVEID-AFCMOLECULAR FORMULA: C38H44F3N5O12MOLECULAR WEIGHT:819.8STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [219138-06-4], RESEARCH AREA: ApoptosisREFERENCES: N.A. Thornberry, and Y. Lazebnik, Science 281, 1312 (1998); R.V. Talanian, et al., J. Biol. Chem. 272, 9677 (1997)
Catalog No. 50-210-9056