Learn More
AdipoGen Z-Val-Leu-Arg-AMC
SDP
Catalog No. 502562431
Encompass_Preferred
Chemical. Formula C35H47N7O7 . C2HF3O2. MW 677.8 . 114.0. Proteasome subunit alpha type-1 Macropain substrate used for assaying the trypsin-like activity of the 20S proteasome. Cysteine proteinase falcipain-2 and falcipain-3 malarial hemoglobinases substrate. Cathepsin S substrate. Hydrolysis of this fluorogenic peptide substrate is monitored by observing fluorescence at an Excitation of 345nm and Emission at 445nm.
Catalog No. 50-256-2431 Supplier AdipoGen Supplier No. AGCP30027M025
Chemical. Formula C35H47N7O7 . C2HF3O2. MW 677.8 . 114.0. Proteasome subunit alpha type-1 Macropain substrate used for assaying the trypsin-like activity of the 20S proteasome. Cysteine proteinase falcipain-2 and falcipain-3 malarial hemoglobinases substrate. Cathepsin S substrate. Hydrolysis of this fluorogenic peptide substrate is monitored by observing fluorescence at an Excitation of 345nm and Emission at 445nm.