Learn More
Medchemexpress LLC Z-Val-Lys-Met-AMC (TFA) | 141223-71-4 | 99.9% | 667.82 | 1 MG
SDP
Catalog No. 5000398745
Encompass_Preferred
Z-Val-Lys-Met-AMC (TFA) is a fluorescent substrate utilized for detecting β-secretase activity of cathepsin B.
  • Fluorescent substrate for cathepsin B activity
  • Appears as a white to off-white solid
  • Store as powder at -80°C for up to 2 years
  • Store in solvent at -80°C for 6 months, away from moisture
  • Shipping at room temperature is suitable for durations less than 2 weeks
  • For laboratory research use only
Catalog No. 50-003-98745 Supplier Medchemexpress LLC Supplier No. HYP4428A1MG
Z-Val-Lys-Met-AMC (TFA) is a fluorescent substrate utilized for detecting β-secretase activity of cathepsin B.
  • Fluorescent substrate for cathepsin B activity
  • Appears as a white to off-white solid
  • Store as powder at -80°C for up to 2 years
  • Store in solvent at -80°C for 6 months, away from moisture
  • Shipping at room temperature is suitable for durations less than 2 weeks
  • For laboratory research use only