Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Z-val-val-arg-AMC | 124485-41-2 | MFCD00238523 | 99.8% | 663.76 | C34H45N7O7 | 5 MG
SDP

Catalog No. 5000221793
Encompass_Preferred

Z-Val-Val-Arg-AMC is a fluorogenic peptide substrate used to measure cathepsin protease activity in biochemical and enzymatic assays. The peptide contains a benzyloxycarbonyl (Z) protecting group and a 7-amino-4-methylcoumarin (AMC) fluorescent leaving group; cleavage by cathepsins releases AMC and generates a measurable fluorescence signal. For research use only.

  • Fluorogenic substrate for cathepsin activity assays.
  • Sequence: {Z}-Val-Val-Arg-{AMC}.
  • High purity (99.8%).
  • Molecular weight 663.76.
  • Chemical formula C34H45N7O7.
  • Available in small mg-scale packages for assay development.

Catalog No. 50-002-21793 Supplier Medchemexpress LLC Supplier No. HY1373675MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z-Val-Val-Arg-AMC is a fluorogenic peptide substrate used to measure cathepsin protease activity in biochemical and enzymatic assays. The peptide contains a benzyloxycarbonyl (Z) protecting group and a 7-amino-4-methylcoumarin (AMC) fluorescent leaving group; cleavage by cathepsins releases AMC and generates a measurable fluorescence signal. For research use only.

  • Fluorogenic substrate for cathepsin activity assays.
  • Sequence: {Z}-Val-Val-Arg-{AMC}.
  • High purity (99.8%).
  • Molecular weight 663.76.
  • Chemical formula C34H45N7O7.
  • Available in small mg-scale packages for assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.