Learn More
Enzo Life Sciences Z-VE(OMe)ID(OMe)-FMK (1mg)
SDP
Catalog No. 502008289
Encompass_Preferred
Caspase-6 inhibitor. Irreversible, cell permeable inhibitor of caspase-6. Alternative name: Caspase 6 inhibitor (fluoromethylketone). Formula: C31H45FN4O10. MW: 652.9. Purity: ≥95% (TLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO. Long Term Storage: -20°C. Use/Stability: Store stock solutions in aliquots at -20°C. Handling: Packaged under inert gas.
Catalog No. 50-200-8289 Supplier Enzo Life Sciences Supplier No. ALX260075M001
Caspase-6 inhibitor. Irreversible, cell permeable inhibitor of caspase-6. Alternative name: Caspase 6 inhibitor (fluoromethylketone). Formula: C31H45FN4O10. MW: 652.9. Purity: ≥95% (TLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO. Long Term Storage: -20°C. Use/Stability: Store stock solutions in aliquots at -20°C. Handling: Packaged under inert gas.