Learn More
Cayman Chemical Z-VEID-AFC (trifluoroacetate salt), Purity: ≥95%, 1 mg
SDP
Catalog No. 502939556
Encompass_Preferred
A fluorogenic substrate for caspase-6; AFC is released upon enzymatic cleavage by caspase-6 and its fluorescence can be used to quantify caspase-6 activity; ex/em = 400/505 nm, respectively
Catalog No. 50-293-9556 Supplier Cayman Chemical Supplier No. 271461 MG
A fluorogenic substrate for caspase-6; AFC is released upon enzymatic cleavage by caspase-6 and its fluorescence can be used to quantify caspase-6 activity; ex/em = 400/505 nm, respectively