Learn More
Apexbio Technology LLC Z-VEID-FMK,1mg
SDP
Supplier:  Apexbio Technology LLC A19231
Encompass_Preferred
When compared to other caspase inhibitors, Z-DRHD-FMK inhibited caspase 6 activity more effectively than the general caspase inhibitor Z-Val-Ala-Asp (OMe)-fluoromethy ketone (Z-VAD-FMK) or the caspase 6 inhibitor Z-Val-Glu(Ome)-Ile-Asp(OMe)-fluoromethyl ketone (Z-VEID-FMK). However, it was less effective in inhibiting TNFα-induced apoptosis than Z-VAD-FMK or Z-VEID-FMK, presumably because caspase 6 is only one of at least three effector caspases, the others being caspase 3 and 7, that are active during caspase dependent apoptosis. Loss of DNA-binding activity and TNFα-induced apoptosis can be prevented by caspase-6-preferred inhibitor (Z-VEID-FMK)1. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-223-9987