Learn More
Abcam Z-VEID-FMK, caspase-6 inhibitor, 20UL
Catalog No. 502706373
Encompass_Preferred
MW 652.7 Da, Purity >98%. Z-VEID-FMK, caspase-6 inhibitor. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6373 Supplier Abcam Supplier No. AB14202520UL
MW 652.7 Da, Purity >98%. Z-VEID-FMK, caspase-6 inhibitor. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement