Learn More
Enzo Life Sciences Z-VEID-FMK (Ready-to-Use) (100µl)
SDP
Supplier:  Enzo Life Sciences ALX260143R100
Encompass_Preferred
Irreversible, cell permeable inhibitor of caspase-6. The inhibitor is designed as a methyl ester to facilitate cell permeability. Alternative name: Caspase 6 inhibitor (fluoromethylketone). MW: 652.0. Purity: ≥98% (HPLC). Formulation: Liquid. In DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8351