Learn More
Enzo Life Sciences Z-VRPR-FMK (0.5mg)
SDP
Catalog No. 502008364
Encompass_Preferred
MALT1 inhibitor. The proteolytic activity of the paracaspase MALT1 is key to NF-κB and T cell activation. Z-VRPR-FMK is a selective MALT1 inhibitor of such proteolytic activity. Alternative name: Z-Val-Arg-Pro-DL-Arg-FMK, MALT1 inhibitor. Formula: C31H49FN10O6 . MW: 676.8 . 114. Purity: ≥90% (HPLC). Solubility: Soluble in water (1mg/ml). Long Term Storage: -20°C. Handling: Hygroscopic. Avoid freeze/thaw cycles.
Catalog No. 50-200-8364 Supplier Enzo Life Sciences Supplier No. ALX260166MC05
MALT1 inhibitor. The proteolytic activity of the paracaspase MALT1 is key to NF-κB and T cell activation. Z-VRPR-FMK is a selective MALT1 inhibitor of such proteolytic activity. Alternative name: Z-Val-Arg-Pro-DL-Arg-FMK, MALT1 inhibitor. Formula: C31H49FN10O6 . MW: 676.8 . 114. Purity: ≥90% (HPLC). Solubility: Soluble in water (1mg/ml). Long Term Storage: -20°C. Handling: Hygroscopic. Avoid freeze/thaw cycles.