Learn More
AdipoGen Z-VRPR-FMK
SDP
Supplier:  AdipoGen AGCP30008M001
Encompass_Preferred
Chemical. CAS 1381885-28-4 free base. Formula C31H49FN10O6 . C2HF3O2. MW 676.8 . 114.0. Synthetic. Selective cell permeable and irreversible MALT1 inhibitor. Inhibits NF-kappaB-dependent gene expression and affects the growth, proliferation and survival of activated B cell-like diffuse large B cell lymphomas ABC-DLBCL and germinal center B cell like diffuse large B cell lymphoma GCB-DLBCL cell lines. Shown to inhibit the autoprocessing activity of MCA2ac calcium dependent in Trypanosoma brucei.
Catalog No. 50-256-2456