Learn More
Medchemexpress LLC Z-VRPR-FMK TFA | 1381885-28-4 | 95.9% | 790.81 | 500 UG
SDP
Catalog No. 5000424533
Encompass_Preferred
Z-VRPR-FMK (TFA), also known as VRPR, is a tetrapeptide that functions as a selective and irreversible inhibitor of MALT1 (Mucosa-associated lymphoid tissue lymphoma translocation protein 1). It can also protect against influenza A virus (IAV) infection.
  • Functions as a selective and irreversible MALT1 inhibitor.
  • Protects against influenza A virus (IAV) infection.
Catalog No. 50-004-24533 Supplier Medchemexpress LLC Supplier No. HYP1407500UG
Z-VRPR-FMK (TFA), also known as VRPR, is a tetrapeptide that functions as a selective and irreversible inhibitor of MALT1 (Mucosa-associated lymphoid tissue lymphoma translocation protein 1). It can also protect against influenza A virus (IAV) infection.
  • Functions as a selective and irreversible MALT1 inhibitor.
  • Protects against influenza A virus (IAV) infection.