Learn More
Medchemexpress LLC Z-VRPR-FMK TFA | 1926163-57-6 | MFCD18782585 | 95.9% | 790.81 | 100 UG
SDP
Catalog No. 5000423265
Encompass_Preferred
Z-VRPR-FMK (TFA) (VRPR), a tetrapeptide, is a selective and irreversible MALT1 (Mucosa-associated lymphoid tissue lymphoma translocation protein 1) inhibitor. It can protect against influenza A virus (IAV) infection.
  • Selective and irreversible MALT1 inhibitor
  • Tetrapeptide
  • Protects against influenza A virus (IAV) infection
Catalog No. 50-004-23265 Supplier Medchemexpress LLC Supplier No. HYP1407100UG
Z-VRPR-FMK (TFA) (VRPR), a tetrapeptide, is a selective and irreversible MALT1 (Mucosa-associated lymphoid tissue lymphoma translocation protein 1) inhibitor. It can protect against influenza A virus (IAV) infection.
  • Selective and irreversible MALT1 inhibitor
  • Tetrapeptide
  • Protects against influenza A virus (IAV) infection