Learn More
Medchemexpress LLC Z-VRPR-FMK TFA (VRPR) | 95.0% | 790.81 | 200 UG
SDP
Catalog No. 5000423085
Encompass_Preferred
Z-VRPR-FMK TFA (VRPR) is a tetrapeptide and a selective, irreversible MALT1 (Mucosa-associated lymphoid tissue lymphoma translocation protein 1) inhibitor. It can protect against influenza A virus (IAV) infection.
  • Selective and irreversible MALT1 inhibitor
  • Protects against influenza A virus (IAV) infection
Catalog No. 50-004-23085 Supplier Medchemexpress LLC Supplier No. HYP1407200UG
Z-VRPR-FMK TFA (VRPR) is a tetrapeptide and a selective, irreversible MALT1 (Mucosa-associated lymphoid tissue lymphoma translocation protein 1) inhibitor. It can protect against influenza A virus (IAV) infection.
  • Selective and irreversible MALT1 inhibitor
  • Protects against influenza A virus (IAV) infection