Learn More
Abcam Z-VRPR-FMK (Z-Val-Arg-Pro-DL-Arg-FMK, MALT1 inhibitor), MALT1 inhibitor, 1MG
Catalog No. 502705815
Encompass_Preferred
MW 676.79 g/mol, Purity >94%. Selective, cell-permeable and irreversible MALT1 inhibitor. Inhibits NF-κB-dependent gene expression. Shows antiproliferative effects to B cells.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5815 Supplier Abcam Supplier No. AB1443901MG
MW 676.79 g/mol, Purity >94%. Selective, cell-permeable and irreversible MALT1 inhibitor. Inhibits NF-κB-dependent gene expression. Shows antiproliferative effects to B cells.
The product is subject to the following: Abcam Restricted Use Statement