Learn More
Enzo Life Sciences Z-VVR-AMC (10mg). CAS: 124485-41-2
SDP
Catalog No. 502011276
Encompass_Preferred
Sensitive fluorogenic substrate for cathepsin S and cathepsin L. Also cleaved by cathepsin B, cathepsin V, kallikrein 8, and factor VIIa/TF but not cathepsin K nor kallikrein 2. Ex.: 380 nm, Em.: 460 nm, although the following Ex/Em can also be used: 355,375/440,450. This substrate is useful for inhibitor screening and kinetic analysis. Also available: fluorogenic calibration standard AMC (BML-KI144). Alternative name: Cathepsin S & L substrate (fluorogenic). Purity: ≥99% (TLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-1276 Supplier Enzo Life Sciences Supplier No. BMLP1990010
Sensitive fluorogenic substrate for cathepsin S and cathepsin L. Also cleaved by cathepsin B, cathepsin V, kallikrein 8, and factor VIIa/TF but not cathepsin K nor kallikrein 2. Ex.: 380 nm, Em.: 460 nm, although the following Ex/Em can also be used: 355,375/440,450. This substrate is useful for inhibitor screening and kinetic analysis. Also available: fluorogenic calibration standard AMC (BML-KI144). Alternative name: Cathepsin S & L substrate (fluorogenic). Purity: ≥99% (TLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.