Learn More
Medchemexpress LLC Z-WEHD-FMK | 210345-00-9 | 98.0% | 763.77 | 10 MG
SDP
Catalog No. 5000425704
Encompass_Preferred
Z-WEHD-FMK is a potent, cell-permeable, and irreversible caspase-1/5 inhibitor. It also exhibits a robust inhibitory effect on cathepsin B activity (IC50=6 μM) and can be used to investigate cells for evidence of apoptosis.
  • Potent, cell-permeable and irreversible caspase-1/5 inhibitor
  • Exhibits robust inhibitory effect on cathepsin B activity
  • Can be used to investigate cells for evidence of apoptosis
  • Elicits near-complete blockage of C. trachomatis-induced cleavage of golgin-84
  • Inhibits E. piscicida-induced ZF4 cells cytotoxicity and pyroptotic morphology
  • Significantly induces a decrease in IL-1β release in bone marrow-derived macrophages
Catalog No. 50-004-25704 Supplier Medchemexpress LLC Supplier No. HYP011110MG
Z-WEHD-FMK is a potent, cell-permeable, and irreversible caspase-1/5 inhibitor. It also exhibits a robust inhibitory effect on cathepsin B activity (IC50=6 μM) and can be used to investigate cells for evidence of apoptosis.
  • Potent, cell-permeable and irreversible caspase-1/5 inhibitor
  • Exhibits robust inhibitory effect on cathepsin B activity
  • Can be used to investigate cells for evidence of apoptosis
  • Elicits near-complete blockage of C. trachomatis-induced cleavage of golgin-84
  • Inhibits E. piscicida-induced ZF4 cells cytotoxicity and pyroptotic morphology
  • Significantly induces a decrease in IL-1β release in bone marrow-derived macrophages