Learn More
Enzo Life Sciences Z-WEHD-FMK (Ready-to-Use) (100µl)
SDP
Catalog No. 502008345
Encompass_Preferred
Irreversible, cell permeable inhibitor of caspase-1 and -5. Inhibits also caspase-4 and-14. Designed as a methyl ester to facilitate cell permeability. Alternative name: Caspase 1 inhibitor (fluoromethylketone), Caspase 5 inhibitor (fluoromethylketone). Formula: C37H42FN7O11. MW: 763.8. Purity: ≥99% (HPLC). Formulation: Liquid. In DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8345 Supplier Enzo Life Sciences Supplier No. ALX260140R100
Irreversible, cell permeable inhibitor of caspase-1 and -5. Inhibits also caspase-4 and-14. Designed as a methyl ester to facilitate cell permeability. Alternative name: Caspase 1 inhibitor (fluoromethylketone), Caspase 5 inhibitor (fluoromethylketone). Formula: C37H42FN7O11. MW: 763.8. Purity: ≥99% (HPLC). Formulation: Liquid. In DMSO. Long Term Storage: -20°C.