Learn More
Medchemexpress LLC Z-YVAD-AFC | 201608-13-1 | 98.85% | 811.76 | 25 MG
SDP
Catalog No. 5000398972
Encompass_Preferred
Z-YVAD-AFC is a fluorescent substrate for Caspase-1, used to detect and measure its activity. It has a high purity of 98.85% and a molecular weight of 811.76. This product is provided as a solid for research use only.
  • Fluorescent substrate for Caspase-1
  • Detects and measures Caspase-1 activity
  • High purity
  • Available in various quantities
  • For research use only
Catalog No. 50-003-98972 Supplier Medchemexpress LLC Supplier No. 25MGHYP4429
Z-YVAD-AFC is a fluorescent substrate for Caspase-1, used to detect and measure its activity. It has a high purity of 98.85% and a molecular weight of 811.76. This product is provided as a solid for research use only.
  • Fluorescent substrate for Caspase-1
  • Detects and measures Caspase-1 activity
  • High purity
  • Available in various quantities
  • For research use only