Learn More
Cayman Chemical Z-YVAD-AFC (trifluoroacetate salt), Purity: ≥95%, 1 mg
SDP
Catalog No. 502939564
Encompass_Preferred
A fluorogenic substrate for caspase-1; AFC is released upon enzymatic cleavage by caspase-1 and its fluorescence can be used to quantify caspase-1 activity; ex/em = 400/505 nm, respectively
Catalog No. 50-293-9564 Supplier Cayman Chemical Supplier No. 271371 MG
A fluorogenic substrate for caspase-1; AFC is released upon enzymatic cleavage by caspase-1 and its fluorescence can be used to quantify caspase-1 activity; ex/em = 400/505 nm, respectively